Product Details

SNP ID
rs2848444
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60700006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATTAGAATGTATAAGTAGCCTA[A/G]AAAATTGAGCAGTGGAAGCCACCCA
Phenotype
MIM: 606549
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MS4A8 PubMed Links

Gene Details

Gene
MS4A8
Gene Name
membrane spanning 4-domains A8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031457.1 Intron NP_113645.1

View Full Product Details