Product Details

SNP ID
hCV16131667
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105970581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAGATTTATGTCATAGCAGACCT[C/T]GGGAGCATCCTCAGCACTGACACCA
Phenotype
MIM: 146910 MIM: 147070 MIM: 147010
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ADAM6 PubMed Links

Gene Details

Gene
ADAM6
Gene Name
ADAM metallopeptidase domain 6 (pseudogene)
There are no transcripts associated with this gene.

Gene
IGH
Gene Name
immunoglobulin heavy locus
There are no transcripts associated with this gene.

View Full Product Details