Product Details
- SNP ID
-
rs2138615
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:33498870 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTTGAAGTAACAGCTTGTGCCCGA[A/G]AAACTTAACAGATGAGTTCTTGAAT
- Phenotype
-
MIM: 614426
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MAK16
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76212197] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MAK16
- Gene Name
- MAK16 homolog
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032509.3 |
2402 |
UTR 3 |
|
|
NP_115898.2 |
- Gene
- TTI2
- Gene Name
- TELO2 interacting protein 2
View Full Product Details