Product Details

SNP ID
rs2242507
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:50133031 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAAGGAGGTGAGGAAAGAGTGGA[A/G]AGTACAGGGTCTAATCCTTCCTGGA
Phenotype
MIM: 615335
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CERS5 PubMed Links

Gene Details

Gene
CERS5
Gene Name
ceramide synthase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281731.1 988 Intron NP_001268660.1
NM_147190.3 988 Intron NP_671723.1
XM_005269220.2 988 Intron XP_005269277.1
XM_006719687.3 988 Intron XP_006719750.1
XM_011538975.2 988 Missense Mutation CTC,TTC L355F XP_011537277.1
XM_017020202.1 988 Intron XP_016875691.1
XM_017020203.1 988 Missense Mutation CTC,TTC L297F XP_016875692.1
XM_017020204.1 988 Intron XP_016875693.1
XM_017020205.1 988 Missense Mutation CTC,TTC L211F XP_016875694.1

View Full Product Details