Product Details

SNP ID
rs2272936
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:6635917 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
TTCCAACGCGCTCACTCTAATTCTA[C/T]CTGTGAAAGGGCCCCAGCCCCCCTT
Phenotype
MIM: 614827 MIM: 612532
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC11 PubMed Links

Gene Details

Gene
DNAJC11
Gene Name
DnaJ heat shock protein family (Hsp40) member C11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018198.3 Intron NP_060668.2
XM_017001745.1 Intron XP_016857234.1
Gene
THAP3
Gene Name
THAP domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195752.1 Intron NP_001182681.1
NM_001195753.1 Intron NP_001182682.1
NM_138350.3 Intron NP_612359.2
XM_005263532.3 Intron XP_005263589.1
XM_011542400.2 Intron XP_011540702.1
XM_011542401.2 Intron XP_011540703.1
XM_017002761.1 Intron XP_016858250.1

View Full Product Details