Product Details

SNP ID
rs2272992
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:40373862 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
GTGGTCCGCCTTTGCCTGGGCTGAG[A/G]GTCTCTGGCCCCGCAGCCTCTCTTG
Phenotype
MIM: 616916
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMAP2 PubMed Links

Gene Details

Gene
SMAP2
Gene Name
small ArfGAP2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198978.1 157 Intron NP_001185907.1
NM_001198979.1 157 Intron NP_001185908.1
NM_001198980.1 157 Intron NP_001185909.1
NM_022733.2 157 UTR 5 NP_073570.1
XM_011541960.1 157 UTR 5 XP_011540262.1
XM_017002093.1 157 UTR 5 XP_016857582.1

View Full Product Details