Product Details

SNP ID
rs2273439
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:101979364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCGTGATTGATGCAGATAAACC[C/T]GTGTCTTCTCAGCTCCGGGTCCTTA
Phenotype
MIM: 600112
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DYNC1H1 PubMed Links

Gene Details

Gene
DYNC1H1
Gene Name
dynein cytoplasmic 1 heavy chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001376.4 554 Silent Mutation CCC,CCT P130P NP_001367.2

View Full Product Details