Product Details

SNP ID
rs2297843
Assay Type
Validated
NCBI dbSNP Submissions
25
Location
Chr.1:9943399 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
CGCCGGGGGAGCACTTCCGCTGGAC[C/G]CACGCACTTCCGGCCCCCTGGAACC
Phenotype
MIM: 610458 MIM: 608700
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LZIC PubMed Links

Gene Details

Gene
LZIC
Gene Name
leucine zipper and CTNNBIP1 domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316973.1 29 UTR 5 NP_001303902.1
NM_001316974.1 29 UTR 5 NP_001303903.1
NM_001316975.1 29 UTR 5 NP_001303904.1
NM_001316976.1 29 UTR 5 NP_001303905.1
NM_032368.4 29 UTR 5 NP_115744.2
XM_005263506.2 29 Intron XP_005263563.1
XM_005263509.2 29 Intron XP_005263566.1
XM_017002546.1 29 UTR 5 XP_016858035.1
XM_017002547.1 29 Intron XP_016858036.1
Gene
NMNAT1
Gene Name
nicotinamide nucleotide adenylyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297778.1 29 Intron NP_001284707.1
NM_001297779.1 29 Intron NP_001284708.1
NM_022787.3 29 Intron NP_073624.2
XM_011541971.2 29 Intron XP_011540273.1
XM_017002107.1 29 Intron XP_016857596.1
XM_017002108.1 29 Intron XP_016857597.1

View Full Product Details