Product Details

SNP ID
rs2230678
Assay Type
Functionally tested
NCBI dbSNP Submissions
66
Location
Chr.1:29148829 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTCCTCTCTGCTTCTCTTCCTGC[T/C]CTTCCTCTTGTCCTTGCTCTTGCTG
Phenotype
MIM: 601940
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
SRSF4 PubMed Links

Gene Details

Gene
SRSF4
Gene Name
serine and arginine rich splicing factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005626.4 1149 Missense Mutation AGC,GGC S356G NP_005617.2
XM_011541951.2 1149 Missense Mutation AGC,GGC S326G XP_011540253.1

View Full Product Details