Product Details

SNP ID
rs2995308
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133266358 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGCCAGGCGAAGGGCATGCAGGA[C/T]TCTCTAGGCTATTTTTGCAACTTTT
Phenotype
MIM: 602267
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ADAM8 PubMed Links
Additional Information
For this assay, SNP(s) [rs77255558] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ADAM8
Gene Name
ADAM metallopeptidase domain 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109.4 Intron NP_001100.3
NM_001164489.1 Intron NP_001157961.1
NM_001164490.1 Intron NP_001157962.1
XM_011539117.2 Intron XP_011537419.2
XM_017015465.1 Intron XP_016870954.1
XM_017015466.1 Intron XP_016870955.1

View Full Product Details