Product Details
- SNP ID
-
rs2395075
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:74008422 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAAATTCATAATGTGAGAGTATGTT[A/G]CCTTCTATCTGAAAGTGGCGTTGGG
- Phenotype
-
MIM: 193065
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
VCL
PubMed Links
Gene Details
- Gene
- VCL
- Gene Name
- vinculin
View Full Product Details