Product Details

SNP ID
rs2425680
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44917802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTACTCAGAGGAGGGTCCAGTCAA[C/T]CTGTCAAACAAATCTTGGTGGGCTT
Phenotype
MIM: 601289
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PABPC1L PubMed Links

Gene Details

Gene
PABPC1L
Gene Name
poly(A) binding protein cytoplasmic 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001124756.2 Intron NP_001118228.1
Gene
YWHAB
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta
There are no transcripts associated with this gene.

View Full Product Details