Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001322026.1 | Intron | NP_001308955.1 | |||
NM_001322027.1 | Intron | NP_001308956.1 | |||
NM_001322028.1 | Intron | NP_001308957.1 | |||
NM_001322029.1 | Intron | NP_001308958.1 | |||
NM_001322030.1 | Intron | NP_001308959.1 | |||
NM_001322031.1 | Intron | NP_001308960.1 | |||
NM_001322032.1 | Intron | NP_001308961.1 | |||
NM_017926.3 | Intron | NP_060396.2 | |||
NM_017972.3 | Intron | NP_060442.2 | |||
XM_006720191.2 | Intron | XP_006720254.1 | |||
XM_017021427.1 | Intron | XP_016876916.1 | |||
XM_017021428.1 | Intron | XP_016876917.1 | |||
XM_017021429.1 | Intron | XP_016876918.1 | |||
XM_017021430.1 | Intron | XP_016876919.1 | |||
XM_017021431.1 | Intron | XP_016876920.1 | |||
XM_017021432.1 | Intron | XP_016876921.1 | |||
XM_017021433.1 | Intron | XP_016876922.1 |