Product Details
- SNP ID
-
rs2601788
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:3957085 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TACTGTCTAAGCCTGACATTAAACT[C/T]ACACCCTCCCCCTCCATCTGGGTTC
- Phenotype
-
MIM: 603302
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADCY9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78377720] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADCY9
- Gene Name
- adenylate cyclase 9
- Gene
- LOC102724927
- Gene Name
- uncharacterized LOC102724927
There are no transcripts associated with this gene.
View Full Product Details