Product Details
- SNP ID
-
rs2584246
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:39371997 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAATCAATAGAGGGTCATGTGGCCC[A/C]TGTCTTGTTTAGCTTCTGAGCATCA
- Phenotype
-
MIM: 120940
MIM: 601236
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs371411060] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C9
- Gene Name
- complement component 9
There are no transcripts associated with this gene.
- Gene
- DAB2
- Gene Name
- DAB2, clathrin adaptor protein
View Full Product Details