Product Details
- SNP ID
-
rs2661874
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:33265536 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCACATCAGGACTCAGCTTGATATC[A/C]TAGGTCAACATATCTGTGCCTTCGT
- Phenotype
-
MIM: 601497
MIM: 610900
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
BAG1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111809650] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BAG1
- Gene Name
- BCL2 associated athanogene 1
- Gene
- CHMP5
- Gene Name
- charged multivesicular body protein 5
View Full Product Details