Product Details

SNP ID
rs2640595
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57726854 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGGCGCCCACACCCGGCGCCG[C/G]CTCCCCCACATGGCCAAGCCTACTT
Phenotype
MIM: 605476 MIM: 609677
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AGAP2 PubMed Links

Gene Details

Gene
AGAP2
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122772.2 Intron NP_001116244.1
NM_014770.3 Intron NP_055585.1
XM_005268625.3 Intron XP_005268682.1
XM_005268626.2 Intron XP_005268683.1
XM_017018772.1 Intron XP_016874261.1
Gene
AGAP2-AS1
Gene Name
AGAP2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
OS9
Gene Name
OS9, endoplasmic reticulum lectin
There are no transcripts associated with this gene.

View Full Product Details