Product Details

SNP ID
rs2231605
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3012552 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTCGGCTGGGGGAGAGCAGAGGC[A/G]GCTGGGGAGGGGCTGCGGAAGGAGG
Phenotype
MIM: 615798 MIM: 615799 MIM: 605914
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CLDN6 PubMed Links

Gene Details

Gene
CLDN6
Gene Name
claudin 6
There are no transcripts associated with this gene.

Gene
CLDN9
Gene Name
claudin 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020982.3 97 UTR 5 NP_066192.1
Gene
TNFRSF12A
Gene Name
TNF receptor superfamily member 12A
There are no transcripts associated with this gene.

View Full Product Details