Product Details

SNP ID
rs73033922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:306568 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCAAGTGAGTTTGCTGCAACTGA[C/T]CTTTTGCTGCTTGACCGTTCCTCTC
Phenotype
MIM: 606517 MIM: 601057
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AHRR PubMed Links

Gene Details

Gene
AHRR
Gene Name
aryl-hydrocarbon receptor repressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242412.1 Intron NP_001229341.1
NM_020731.4 Intron NP_065782.2
Gene
PDCD6
Gene Name
programmed cell death 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267556.1 Intron NP_001254485.1
NM_001267557.1 Intron NP_001254486.1
NM_001267558.1 Intron NP_001254487.1
NM_001267559.1 Intron NP_001254488.1
NM_013232.3 Intron NP_037364.1

View Full Product Details