Product Details

SNP ID
rs140803474
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:24357379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCAACCACTGAGGTGTGGGGGG[A/G]TAGGGATCTGCATTTCTTCATATCA
Phenotype
MIM: 605755 MIM: 608211
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCDC2 PubMed Links

Gene Details

Gene
DCDC2
Gene Name
doublecortin domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195610.1 477 Intron NP_001182539.1
NM_016356.4 477 Intron NP_057440.2
Gene
KAAG1
Gene Name
kidney associated antigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181337.3 477 UTR 5 NP_851854.1

View Full Product Details