Product Details

SNP ID
rs28461223
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:43876600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGCACCAGAGCACAGCCCCACA[C/T]GGGTGCCCCATCAGACAGGGCTGCC
Phenotype
MIM: 611986 MIM: 610337
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MRPS24 PubMed Links
Additional Information
For this assay, SNP(s) [rs10624354] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MRPS24
Gene Name
mitochondrial ribosomal protein S24
There are no transcripts associated with this gene.

Gene
URGCP
Gene Name
upregulator of cell proliferation
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077663.2 3352 UTR 3 NP_001071131.1
NM_001077664.2 3352 UTR 3 NP_001071132.1
NM_001290075.1 3352 UTR 3 NP_001277004.1
NM_001290076.1 3352 UTR 3 NP_001277005.1
NM_017920.4 3352 UTR 3 NP_060390.3
Gene
URGCP-MRPS24
Gene Name
URGCP-MRPS24 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204871.1 3352 Intron NP_001191800.1

View Full Product Details