Product Details

SNP ID
rs35635747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:32529655 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGCCTCCTGGGACTCTTGGTGA[A/G]GCATCCTATGTAAAAATAGGAAAAT
Phenotype
MIM: 615890
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DYNC1LI1 PubMed Links

Gene Details

Gene
DYNC1LI1
Gene Name
dynein cytoplasmic 1 light intermediate chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016141.3 1295 Silent Mutation NP_057225.2

View Full Product Details