Product Details

SNP ID
rs61734435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:150409702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCGGAGCGTGTCTGGGGCGCTC[C/G]CCAGTACCCTGGCGGCGGCTGCCAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TSC22D2 PubMed Links

Gene Details

Gene
TSC22D2
Gene Name
TSC22 domain family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303264.1 1368 Missense Mutation CCC,GCC P118A NP_001290193.1
NM_014779.3 1368 Missense Mutation CCC,GCC P118A NP_055594.1
XM_011513337.2 1368 Missense Mutation CCC,GCC P118A XP_011511639.1

View Full Product Details