Product Details

SNP ID
rs10137936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74942467 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACGAGGCCACGTGTCTTGCTTCTT[C/T]CAAAGCGGAAGCTCCCAGGGGTCTG
Phenotype
MIM: 601121
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PGF PubMed Links

Gene Details

Gene
PGF
Gene Name
placental growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207012.1 1274 UTR 3 NP_001193941.1
NM_001293643.1 1274 UTR 3 NP_001280572.1
NM_002632.5 1274 UTR 3 NP_002623.2

View Full Product Details