Product Details

SNP ID
rs138872052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:44970289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAAGCCTTGGTGTGCAGGTTGGG[C/T]GGGGGAGTCTTCACATTTGCCATCC
Phenotype
MIM: 611309 MIM: 610559
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf10 PubMed Links

Gene Details

Gene
C10orf10
Gene Name
chromosome 10 open reading frame 10
There are no transcripts associated with this gene.

Gene
RASSF4
Gene Name
Ras association domain family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032023.3 227 Intron NP_114412.2
XM_005271833.4 227 Intron XP_005271890.1
XM_005271835.4 227 Intron XP_005271892.1
XM_006718013.3 227 Intron XP_006718076.1
XM_006718014.3 227 UTR 5 XP_006718077.1
XM_011540255.2 227 Intron XP_011538557.1
XM_017016754.1 227 UTR 5 XP_016872243.1
XM_017016755.1 227 UTR 5 XP_016872244.1
XM_017016756.1 227 Intron XP_016872245.1
XM_017016757.1 227 UTR 5 XP_016872246.1
XM_017016758.1 227 UTR 5 XP_016872247.1

View Full Product Details