Product Details

SNP ID
rs77005706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23756312 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAACCTCCCCCCTCCTCCAAGGT[A/G]GGGGGAAGGAAGGTGGCTGCTGGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHP2 PubMed Links

Gene Details

Gene
CHP2
Gene Name
calcineurin like EF-hand protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022097.3 Intron NP_071380.1

View Full Product Details