Product Details

SNP ID
rs61731898
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.1:85267793 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGATGGTATTTTCAGTTCTGCCT[A/T]CTTCTAGAACAGGCAAATTCAGAGA
Phenotype
MIM: 603517
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
BCL10 PubMed Links

Gene Details

Gene
BCL10
Gene Name
B-cell CLL/lymphoma 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320715.1 957 Missense Mutation GAA,GTA E168V NP_001307644.1
NM_003921.4 957 Missense Mutation GAA,GTA E179V NP_003912.1
XM_011542397.2 957 Missense Mutation GAA,GTA E232V XP_011540699.1
XM_011542398.2 957 Missense Mutation GAA,GTA E221V XP_011540700.1
XM_011542399.2 957 Missense Mutation GAA,GTA E161V XP_011540701.1
Gene
C1orf52
Gene Name
chromosome 1 open reading frame 52
There are no transcripts associated with this gene.

Gene
LOC646626
Gene Name
uncharacterized LOC646626
There are no transcripts associated with this gene.

View Full Product Details