Product Details

SNP ID
rs917438881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:129739552 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGAAGGAACATCAAGTCCCCAG[A/G]TATAGCCACGGCGGCTCTTGGCCCA
Phenotype
MIM: 605172
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTGES PubMed Links

Gene Details

Gene
PTGES
Gene Name
prostaglandin E synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004878.4 571 UTR 3 NP_004869.1

View Full Product Details