Product Details
- SNP ID
-
hCV25616974
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:167054501 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCAGGGGACAGAGGGGAAGGATTT[G/T]CTCTCAGGTGGACCCTCAAGGGAGC
- Phenotype
-
MIM: 602171
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GPA33
PubMed Links
Gene Details
- Gene
- GPA33
- Gene Name
- glycoprotein A33
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005814.2 |
|
Intron |
|
|
NP_005805.1 |
View Full Product Details