Product Details

SNP ID
rs34737311
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17405333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCGGCCTGGGCCTCCACATCCTG[A/G]AAGCCCTTCTGGGCCTCGGTCAGCT
Phenotype
MIM: 600534
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BISPR PubMed Links

Gene Details

Gene
BISPR
Gene Name
BST2 interferon stimulated positive regulator (non-protein coding)
There are no transcripts associated with this gene.

Gene
BST2
Gene Name
bone marrow stromal cell antigen 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004335.3 324 Silent Mutation TTC,TTT F81F NP_004326.1
Gene
LOC105372343
Gene Name
uncharacterized LOC105372343
There are no transcripts associated with this gene.

Gene
MVB12A
Gene Name
multivesicular body subunit 12A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304547.1 324 Intron NP_001291476.1
NM_138401.3 324 Intron NP_612410.1

View Full Product Details