Product Details

SNP ID
hCV25619085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:113979497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACTGTACGTTCCTTCTACTCTG[A/G]CACCACTCTCCAGGCTGCCATGGGG
Phenotype
MIM: 608003 MIM: 610088
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIPK1 PubMed Links

Gene Details

Gene
HIPK1
Gene Name
homeodomain interacting protein kinase 1
There are no transcripts associated with this gene.

Gene
OLFML3
Gene Name
olfactomedin like 3
There are no transcripts associated with this gene.

View Full Product Details