Product Details

SNP ID
rs78916864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58622890 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTAAGTTGCCTATTTTGCTGTTA[C/T]CTGTTTTCCCTTCATCTTTTTTGAT
Phenotype
MIM: 118945
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNTF PubMed Links

Gene Details

Gene
CNTF
Gene Name
ciliary neurotrophic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000614.3 Intron NP_000605.1
Gene
ZFP91
Gene Name
ZFP91 zinc finger protein
There are no transcripts associated with this gene.

Gene
ZFP91-CNTF
Gene Name
ZFP91-CNTF readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details