Product Details

SNP ID
rs34361405
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:33376888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATCCTTGACTATCAAAACTGGTC[G/A]CCCGGCCAGGTAACTGAAAGACAAA
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
SYT10 PubMed Links

Gene Details

Gene
SYT10
Gene Name
synaptotagmin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198992.3 1172 Missense Mutation GCG,GTG A505V NP_945343.1
XM_011520644.2 1172 Missense Mutation GCG,GTG A324V XP_011518946.1

View Full Product Details