Product Details

SNP ID
rs149601211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:177433400 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTCAGAATAAATACTAACTCCA[A/G]ATTGGGTTACTTGAAGTTAGAAATT
Phenotype
MIM: 613228
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGA PubMed Links

Gene Details

Gene
AGA
Gene Name
aspartylglucosaminidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000027.3 Intron NP_000018.2
NM_001171988.1 Intron NP_001165459.1
Gene
LOC285500
Gene Name
uncharacterized LOC285500
There are no transcripts associated with this gene.

View Full Product Details