Product Details

SNP ID
rs62116990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43797456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCAGGTTGTGGGGCACAAGGGC[A/G]GGAGAGATGGAAGGCCAGAGGGACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LYPD5 PubMed Links

Gene Details

Gene
LYPD5
Gene Name
LY6/PLAUR domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031749.2 1022 UTR 3 NP_001026919.2
NM_001288763.1 1022 UTR 3 NP_001275692.1
NM_182573.2 1022 UTR 3 NP_872379.2
XM_011526794.2 1022 UTR 3 XP_011525096.1

View Full Product Details