Product Details

SNP ID
rs34490804
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50182703 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACCGGCGCAGCAATCAAGTGGC[C/T]CGGGCGCTGCACGACCACCTCGGCC
Phenotype
MIM: 609123 MIM: 603247
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP8B4 PubMed Links

Gene Details

Gene
ATP8B4
Gene Name
ATPase phospholipid transporting 8B4 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024837.3 508 Intron NP_079113.2
XM_011522046.2 508 Intron XP_011520348.1
XM_011522047.2 508 Intron XP_011520349.1
XM_011522048.1 508 Intron XP_011520350.1
XM_011522049.2 508 Intron XP_011520351.1
XM_011522051.2 508 Intron XP_011520353.1
XM_011522052.2 508 Intron XP_011520354.1
XM_011522053.1 508 Intron XP_011520355.1
XM_011522056.2 508 Intron XP_011520358.2
XM_011522058.2 508 Intron XP_011520360.1
XM_011522059.1 508 Intron XP_011520361.1
XM_011522060.1 508 Intron XP_011520362.1
XM_011522061.1 508 Intron XP_011520363.1
XM_011522062.1 508 Intron XP_011520364.1
XM_011522063.1 508 Intron XP_011520365.1
XM_011522064.1 508 Intron XP_011520366.1
XM_011522069.2 508 Intron XP_011520371.1
XM_011522070.1 508 Intron XP_011520372.1
XM_017022587.1 508 Intron XP_016878076.1
XM_017022588.1 508 Intron XP_016878077.1
XM_017022589.1 508 Intron XP_016878078.1
XM_017022590.1 508 Intron XP_016878079.1
XM_017022591.1 508 Intron XP_016878080.1
XM_017022592.1 508 Intron XP_016878081.1
XM_017022593.1 508 Intron XP_016878082.1
XM_017022594.1 508 Intron XP_016878083.1
XM_017022595.1 508 Intron XP_016878084.1
XM_017022596.1 508 Intron XP_016878085.1
XM_017022597.1 508 Intron XP_016878086.1
XM_017022598.1 508 Intron XP_016878087.1
XM_017022599.1 508 Intron XP_016878088.1
Gene
SLC27A2
Gene Name
solute carrier family 27 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159629.1 508 Silent Mutation GCC,GCT A92A NP_001153101.1
NM_003645.3 508 Silent Mutation GCC,GCT A92A NP_003636.2

View Full Product Details