Product Details

SNP ID
rs190309436
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:120538591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGTGACAAAAGAGGAAATCGATT[A/G]AAAGGGTAAAAAGCTAACATTCTCC
Phenotype
MIM: 300304
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CUL4B PubMed Links

Gene Details

Gene
CUL4B
Gene Name
cullin 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079872.1 Intron NP_001073341.1
NM_003588.3 Intron NP_003579.3
XM_005262481.1 Intron XP_005262538.1
XM_006724784.1 Intron XP_006724847.1
XM_006724785.2 Intron XP_006724848.1
XM_011531399.2 Intron XP_011529701.1
XM_011531400.2 Intron XP_011529702.1
XM_011531401.1 Intron XP_011529703.1

View Full Product Details