Product Details

SNP ID
rs62642551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:40827224 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTACCAGGCATTCCCCCAGGAA[A/G]GCCACCTGTAATAAAAAATGAATAG
Phenotype
MIM: 606795 MIM: 606796
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC25A17 PubMed Links

Gene Details

Gene
SLC25A17
Gene Name
solute carrier family 25 member 17
There are no transcripts associated with this gene.

Gene
ST13
Gene Name
suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278589.1 1321 Missense Mutation NP_001265518.1
NM_003932.4 1321 Missense Mutation NP_003923.2

View Full Product Details