Product Details

SNP ID
rs74717897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10451182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGCAGTGTGTGATGTCAGGGAC[C/T]GTACTCTATAATAACAGTAGTTAAG
Phenotype
MIM: 161555
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLRC1 PubMed Links

Gene Details

Gene
KLRC1
Gene Name
killer cell lectin like receptor C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304448.1 432 UTR 5 NP_001291377.1
NM_002259.4 432 UTR 5 NP_002250.1
NM_007328.3 432 UTR 5 NP_015567.1
NM_213657.2 432 UTR 5 NP_998822.1
NM_213658.2 432 UTR 5 NP_998823.1

View Full Product Details