Product Details

SNP ID
rs34247288
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31372984 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCATTCCATCCAATGAAGACCA[C/T]AGGCGAGTTCCTGCGACATCTCCCA
Phenotype
MIM: 607650 MIM: 615997
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DEFB118 PubMed Links

Gene Details

Gene
DEFB118
Gene Name
defensin beta 118
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_054112.2 219 Silent Mutation CAC,CAT H62H NP_473453.1
Gene
DEFB119
Gene Name
defensin beta 119
There are no transcripts associated with this gene.

View Full Product Details