Product Details
- SNP ID
-
rs61737277
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:122977658 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCCAGCCTGGCGGCTCAGAGCAC[G/T]TGCGGCCCTGAGCCCAGCTCCCCCT
- Phenotype
-
MIM: 611074
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
BSX
PubMed Links
Gene Details
- Gene
- BSX
- Gene Name
- brain specific homeobox
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001098169.1 |
693 |
Missense Mutation |
CAA,CAC |
Q231H |
NP_001091639.1 |
View Full Product Details