Product Details

SNP ID
rs59586431
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:138076673 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTTTAACATTTGCCTATAGCA[C/T]AATCTGCAATTTATTTTAAGCAGAT
Phenotype
MIM: 604741
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKR1D1 PubMed Links

Gene Details

Gene
AKR1D1
Gene Name
aldo-keto reductase family 1, member D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190906.1 Intron NP_001177835.1
NM_001190907.1 Intron NP_001177836.1
NM_005989.3 Intron NP_005980.1

View Full Product Details