Product Details

SNP ID
rs192250259
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:156244586 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGCTACTAGTTTTGCTACCTGA[G/T]AAGTGGCAGGTGGAGGACCCTTCCA
Phenotype
MIM: 112260 MIM: 614579 MIM: 609176 MIM: 610962
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BGLAP PubMed Links

Gene Details

Gene
BGLAP
Gene Name
bone gamma-carboxyglutamate protein
There are no transcripts associated with this gene.

Gene
PAQR6
Gene Name
progestin and adipoQ receptor family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272104.1 Intron NP_001259033.1
NM_001272105.1 Intron NP_001259034.1
NM_001272106.1 Intron NP_001259035.1
NM_001272107.1 Intron NP_001259036.1
NM_001272108.1 Intron NP_001259037.1
NM_001272109.1 Intron NP_001259038.1
NM_001272110.1 Intron NP_001259039.1
NM_001272111.1 Intron NP_001259040.1
NM_001272112.1 Intron NP_001259041.1
NM_001272113.1 Intron NP_001259042.1
NM_024897.3 Intron NP_079173.2
NM_198406.2 Intron NP_940798.1
XM_005245494.3 Intron XP_005245551.1
XM_006711546.2 Intron XP_006711609.1
XM_006711547.3 Intron XP_006711610.1
XM_006711548.3 Intron XP_006711611.1
XM_006711552.2 Intron XP_006711615.1
XM_006711553.3 Intron XP_006711616.1
XM_011510000.2 Intron XP_011508302.1
XM_017002385.1 Intron XP_016857874.1
Gene
PMF1
Gene Name
polyamine-modulated factor 1
There are no transcripts associated with this gene.

Gene
PMF1-BGLAP
Gene Name
PMF1-BGLAP readthrough
There are no transcripts associated with this gene.

Gene
SMG5
Gene Name
SMG5, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.

View Full Product Details