Product Details

SNP ID
rs61995959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:106860608 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTCCAAAACCCAGCAGTAATAG[A/G]AGCTTTCAACCTGAAATACGATTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105376204 PubMed Links

Gene Details

Gene
LOC105376204
Gene Name
uncharacterized LOC105376204
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017015409.1 2408 UTR 5 XP_016870898.1
Gene
ZNF462
Gene Name
zinc finger protein 462
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021224.4 2408 Intron NP_067047.4
XM_006717209.3 2408 Intron XP_006717272.1
XM_006717211.3 2408 Intron XP_006717274.1
XM_006717212.3 2408 Intron XP_006717275.1
XM_006717215.3 2408 Intron XP_006717278.1
XM_006717216.3 2408 Intron XP_006717279.1
XM_006717218.3 2408 Intron XP_006717281.1
XM_011518892.2 2408 Intron XP_011517194.1
XM_017014996.1 2408 Intron XP_016870485.1
XM_017014997.1 2408 Intron XP_016870486.1
XM_017014998.1 2408 Intron XP_016870487.1
XM_017014999.1 2408 Intron XP_016870488.1

View Full Product Details