Product Details
- SNP ID
-
rs2257495
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:46367543 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTTCCCCAGAAGGGCACTGAGGCC[A/G]TCAATCCAGATGCAGTACTGTGGGG
- Phenotype
-
MIM: 606421
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ELMO2
PubMed Links
Gene Details
- Gene
- ELMO2
- Gene Name
- engulfment and cell motility 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001318253.1 |
2609 |
Silent Mutation |
GAC,GAT |
D572D |
NP_001305182.1 |
NM_133171.4 |
2609 |
Silent Mutation |
GAC,GAT |
D660D |
NP_573403.1 |
NM_182764.2 |
2609 |
Silent Mutation |
GAC,GAT |
D660D |
NP_877496.1 |
XM_005260496.3 |
2609 |
Silent Mutation |
GAC,GAT |
D660D |
XP_005260553.1 |
XM_005260498.3 |
2609 |
Silent Mutation |
GAC,GAT |
D572D |
XP_005260555.1 |
XM_005260499.3 |
2609 |
Silent Mutation |
GAC,GAT |
D572D |
XP_005260556.1 |
XM_005260500.3 |
2609 |
Silent Mutation |
GAC,GAT |
D572D |
XP_005260557.1 |
XM_005260501.4 |
2609 |
Silent Mutation |
GAC,GAT |
D477D |
XP_005260558.1 |
XM_006723854.3 |
2609 |
Silent Mutation |
GAC,GAT |
D660D |
XP_006723917.1 |
XM_017028009.1 |
2609 |
Silent Mutation |
GAC,GAT |
D477D |
XP_016883498.1 |
XM_017028010.1 |
2609 |
Silent Mutation |
GAC,GAT |
D477D |
XP_016883499.1 |
XM_017028011.1 |
2609 |
Silent Mutation |
GAC,GAT |
D477D |
XP_016883500.1 |
- Gene
- SLC35C2
- Gene Name
- solute carrier family 35 member C2
There are no transcripts associated with this gene.
View Full Product Details