Product Details

SNP ID
rs76668705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48321092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAAGTCTGGAAGACAAAAGCAC[A/G]TACAGAAAAGTTGCTTCTCTGTCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR2115 PubMed Links

Gene Details

Gene
MIR2115
Gene Name
microRNA 2115
There are no transcripts associated with this gene.

Gene
SPINK8
Gene Name
serine peptidase inhibitor, Kazal type 8 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080525.1 Intron NP_001073994.1
XM_017007046.1 Intron XP_016862535.1
XM_017007047.1 Intron XP_016862536.1

View Full Product Details