Product Details
- SNP ID
-
hCV25743045
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:70463183 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTGTCTCAGGAAGCCCCTCCGCT[T/G]GGCCAGAATGGAGCAGCCGAAGGGA
- Phenotype
-
MIM: 608675
- Polymorphism
- T/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FUK
PubMed Links
Gene Details
- Gene
- FUK
- Gene Name
- fucokinase
There are no transcripts associated with this gene.
View Full Product Details