Product Details

SNP ID
rs34071835
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:201900185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCTTGGCTTCCTTTTCATGTAA[A/G]GGTTCATTCTTCTTGACTTTTTCAT
Phenotype
MIM: 602715
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LMOD1 PubMed Links

Gene Details

Gene
LMOD1
Gene Name
leiomodin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012134.2 1076 Silent Mutation CCC,CCT P276P NP_036266.2
Gene
SHISA4
Gene Name
shisa family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198149.2 1076 Intron NP_937792.2
XM_017000419.1 1076 Intron XP_016855908.1

View Full Product Details