Product Details

SNP ID
rs62639688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55913673 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTAAAATAAAATTTTATTTTTCA[C/G]TTTTTTCAGGGCCTCTTTCACATCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR5W2 PubMed Links

Gene Details

Gene
OR5W2
Gene Name
olfactory receptor family 5 subfamily W member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001960.1 910 Missense Mutation CTG,GTG L304V NP_001001960.1

View Full Product Details