Product Details

SNP ID
rs55963099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29996528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGGAGGACCGTTGGCCCAGCGCA[T/C]CCCTTGCCCGTGATACAGGAGCACG
Phenotype
MIM: 604567 MIM: 603365 MIM: 613199
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
DOC2A PubMed Links

Gene Details

Gene
DOC2A
Gene Name
double C2 domain alpha
There are no transcripts associated with this gene.

Gene
HIRIP3
Gene Name
HIRA interacting protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197323.1 Intron NP_001184252.1
NM_003609.4 Intron NP_003600.2
Gene
INO80E
Gene Name
INO80 complex subunit E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304562.1 Intron NP_001291491.1
NM_001304563.1 Intron NP_001291492.1
NM_173618.2 Intron NP_775889.1
XM_011545809.2 Intron XP_011544111.1
XM_011545811.2 Intron XP_011544113.1
XM_011545812.2 Intron XP_011544114.1
XM_017023169.1 Intron XP_016878658.1
Gene
TAOK2
Gene Name
TAO kinase 2
There are no transcripts associated with this gene.

View Full Product Details